Order Entry
Belgium
Orders LinkContactUsLinkComponent
Needles, Spinocan®

Needles, Spinocan®

Needles, Spinocan®, 22, 40 mm, 0,70 mm, Black
Catalog # BRAU4507401-13
Supplier:   B. Braun
Needles, Spinocan®
Catalog # BRAU4507401-13
Supplier:  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 22, 40 mm, 0,70 mm, Black

Price
136,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Colour code:
    Black
  • Ø:
    0,70 mm
  • Gauge:
    22
  • Description:
    Needles, Spinocan®
  • Length:
    40 mm
  • Seq:
    006
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.