Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Needles, Spinocan®

Needles, Spinocan®, 26, 88 mm, 0,45 mm, Brown
  BRAU4502906-01
 :   B. Braun
Needles, Spinocan®
  BRAU4502906-01
 :  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 26, 88 mm, 0,45 mm, Brown

 
136,00 €
   
1 * 25 ST
 
 
  • Colour code:
    Brown
  • Ø:
    0,45 mm
  • Gauge:
    26
  • Description:
    Needles, Spinocan®
  • Length:
    88 mm
  • Seq:
    011
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.