Orders LinkContactUsLinkComponent
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Three-way stopcocks, White
Catalog # BRAU4095146
Supplier:   B. Braun
Multi-way stopcocks, Discofix®
Catalog # BRAU4095146
Supplier:  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Three-way stopcocks, White
Price
141,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Specifications
  • Colour:
    White
  • Description:
    Three-way stopcocks