Shakers
Shaker for four Mojonnier tubes
614-0183
Shakers for determining the fat content in milk according to Mojonnier's method. White PVC housing. Choice of models to accept 4, 6 or 36 Mojonnier extraction tubes. Units provide forceful, uniform and reproducible mixing.
- Timer, 0 to 15 min
- Produce 100 shakes per minute
- Model for 36 butyrometers is supplied with stand
- Description:Shaker for four Mojonnier tubes
Accessories
- Description
- Description
- Description
- Description