Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Funnels, mini

outlet Ø 4 mm, 50 mm, 27 mm
  BOCH8855
 :   BOCHEM
Funnels, mini
  BOCH8855
 :  BOCHEM
Funnels, mini
Funnels, mini

outlet Ø 4 mm, 50 mm, 27 mm

 
5,00 €
   
1 * 1 ST
 

Mini funnel, made of 18/10 stainless steel.

  • Excellent temperature resistance
  • Good chemical resistance
  • Easy to clean
 
  • Funnel O-Ø:
    27 mm
  • Description:
    outlet Ø 4 mm
  • Length:
    50 mm