ContactUsLinkComponent
Fitting for DIN support material d=26,9/12-13mm, cast iron/steel. Combines 26,9mm tubes with 12/13mm DIN rods. Consists in fitting (steel), tube connector M50 (cast iron), screw nut and grommet. null
Catalog # BOCH110
Supplier:  BOCHEM
Fitting for DIN support material d=26,9/12-13mm, cast iron/steel. Combines 26,9mm tubes with 12/13mm DIN rods. Consists in fitting (steel), tube connector M50 (cast iron), screw nut and grommet. null
Catalog # BOCH110
Supplier:  BOCHEM
Frequently Bought Together