8,5 mmZ, 10 mm Schichdicke Qtz Flowcell, 8.5 mm, 10 mm PL, SF, 4x11 ~ For UV and visible wavelength measurements ~ Masked ~ Screw fittings ~ 10 mm Path ~ Use single-cell holder for best results ~ Screw fittings ~ 10 mm Path, 475 µL, 8.5 mmZ, pkg of 1 * 1 items
Catalog # SPET268-808000
Supplier: Thermo Fisher Scientific
Frequently Bought Together