Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Sacks

Storage Sacks

Approx. 500 bags/pack, 0,06 mm, 350×500 mm
Catalog # LAMA121
Supplier:   LAMAPLAST
Storage Sacks
Catalog # LAMA121
Supplier:  LAMAPLAST
Storage Sacks
Storage Sacks

Approx. 500 bags/pack, 0,06 mm, 350×500 mm

Price
244,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

LDPE, neutral storage bags.

  • With bottom weld
  • Can be used for foodstuffs
Specifications
  • Description:
    Approx. 500 bags/pack
  • Dimensions:
    350×500 mm
  • Seq:
    05
  • Thickness:
    0,06 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.