Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade

Agar, Bacteriological Grade

Bottle, 500 g
Catalog # RC-003
Supplier:   G-Biosciences
Agar, Bacteriological Grade
Catalog # RC-003
Supplier:  G-Biosciences
CAS Number:  9002-18-0
Agar, Bacteriological Grade
Agar, Bacteriological Grade

Bottle, 500 g

CAS Number:  9002-18-0
Price
221,00 €
Log in for availability
1 * 500 g
Product Details & Documents

A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.

Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:(C₁₂H₁₈O₉)n
  • MDL Number:MFCD00081288
  • Melting Pt:90 °C
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.