Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bath fluids, Haake

Bath fluids, Haake

Sil 180, –40...+200 °C, >225 °C, 170 °C, 11 cP, Colourless, 10 L
  HAKA9990204
Bath fluids, Haake
  HAKA9990204
 :  
 :  
Bath fluids, Haake

Sil 180, –40...+200 °C, >225 °C, 170 °C, 11 cP, Colourless, 10 L

 :  
 
 
 
Silicone oil 47 V 350 RHODORSIL Plastic bottle 5L 1 * 5 L  83851.360

Le produit 83851.360 est similaire à HAKA9990204, mais le fabricant et les spécifications peuvent varier.

 

These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.

  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency

The oils are not suitable for contact with silicone tubing.

Documents

 

 

  • Pk:
    10 L
  • Colour:
    Colourless
  • Fire point:
    >225 °C
  • Flash point:
    170 °C
  • Temperature range:
    –40...+200 °C
  • Type:
    Sil 180
  • Viscosity:
    11 cP

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.