Flasks, round bottom, with two necks
100 ml, 24/29 NS, 14/23 NS
WITG0.700.100
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one angled side neck (20°). With matt white labelling area.
: DIN 12394
- Capacity:100 ml
- Side necks:14/23 NS
- Centre neck:24/29 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



