Orders LinkContactUsLinkComponent
Brushes, FlexBrush™
Brushes, FlexBrush™
Polyester cover, NovaPoly
Catalog # 141-1016
   Waste Reducing
Supplier:   MICRONOVA
Brushes, FlexBrush™
Catalog # 141-1016
Supplier:  MICRONOVA
   Waste Reducing
Brushes, FlexBrush™
Brushes, FlexBrush™
Polyester cover, NovaPoly
Price
20,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications
  • Description:
    Polyester cover, NovaPoly
  • Seq:
    05
  • Cat. No.:
    141-1016
  • Environmentally Preferable:
    Waste Reducing