Orders LinkContactUsLinkComponent
Padlock control centres
Padlock control centres
Padlock control centre, 8 hooks, Red, 305×50×254 mm
Catalog # BRDY800119
Supplier:   Brady
Padlock control centres
Catalog # BRDY800119
Supplier:  Brady
Padlock control centres
Padlock control centres
Padlock control centre, 8 hooks, Red, 305×50×254 mm
Price
316,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These padlock control centres provide safe storage and controlled access to padlocks.
  • Made from powder-coated steel
  • Each nickel coated metal hook can hold two locks
  • Two keys are provided for cabinet lock
Ordering information: Components not included. Padlocks need to be ordered separately.
Specifications
  • Colour:
    Red
  • Description:
    Padlock control centre, 8 hooks
  • Seq:
    01
  • W×D×H:
    305×50×254 mm