Group Lockout Boxes
Group lockout centre with 8 hooks and 1 slot, Red, 305×64×195 mm, Acrylic
Catalog # BRDY800116
Supplier: Brady
Jump to:
Product Details & Documents
For group lockout situations involving a large number of workers and equipment. Can also be used as a lock storage box.
- Transparent front cover allows for easy viewing of contents
- Hooks for keys and slot for locks
- Components not included
Specifications
- Colour:Red
- Description:Group lockout centre with 8 hooks and 1 slot
- Material:Acrylic
- W×D×H:305×64×195 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



