Orders LinkContactUsLinkComponent
Pipette tips
Standard, 0,5 - 200 µl, +, 51 mm, Yellow, 10 racks, 96 each
Catalog # 732-1557
   Low Manufacturing Impact
Supplier:   Sartorius
Pipette tips
Catalog # 732-1557
Supplier:  Sartorius
   Low Manufacturing Impact
Pipette tips
Pipette tips
Standard, 0,5 - 200 µl, +, 51 mm, Yellow, 10 racks, 96 each
Price
96,80 €
Log in for availability
1 * 960 ST
Product Details & Documents
These pipette tips are suitable for many pipettes on the market.
  • Natural colour, premium grade PP, autoclavable
  • Excellent seal design for accurate, leak-tight sealing
  • Produced and packed in a dust-free environment under strictly maintained conditions
 : Available in bulk packaging, in autoclavable racks and in refills.
 : Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
Specifications
  • Colour code:
    Yellow
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    0,5 - 200 µl
  • Length:
    51 mm
  • Seq:
    10
  • Tip style:
    Standard
  • Cat. No.:
    732-1557
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.