ContactUsLinkComponent
Freezer,gti1853 impulse sales chest, 172l, -10c to -24c, colour white and frame with blue colour, mechanic control system, intern al analo g, swivel castors, 220-240v, 1a, aluminium, manual defrosting, baskets 3. 1 * 1 items
Catalog # LIEBGTI1853
Supplier:  Liebherr
Freezer,gti1853 impulse sales chest, 172l, -10c to -24c, colour white and frame with blue colour, mechanic control system, intern al analo g, swivel castors, 220-240v, 1a, aluminium, manual defrosting, baskets 3. 1 * 1 items
Catalog # LIEBGTI1853
Supplier:  Liebherr
Frequently Bought Together