Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Büchner Funnels

VWR®, Büchner Funnels

334 mm, 10000 ml, 320 mm
  511-0075
VWR®, Büchner Funnels
  511-0075
VWR®, Büchner Funnels
VWR®, Büchner Funnels

334 mm, 10000 ml, 320 mm

 
1 270,00 €
   
1 * 1 ST
 

White glazed porcelain.

For paper or membrane filtration discs.

Documents

 
  • Plate Ø:
    334 mm
  • Capacity:
    10000 ml
  • Seq:
    13
  • For filter Ø:
    320 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.