Tubes and systems for active sampling
Silica gel tubes, B/G
Catalog # DRAE8101861
Supplier: Dräger
Jump to:
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Documents
Specifications
- Type:B/G
- Description:Silica gel tubes
- Seq:21