Order Entry
Belgium
Orders LinkContactUsLinkComponent
Group Lockout Boxes

Group Lockout Boxes

Group lockout centre with 18 hooks and 2 slots, Red, 406×64×195 mm, Acrylic
Catalog # BRDY800127
Supplier:   Brady
Group Lockout Boxes
Catalog # BRDY800127
Supplier:  Brady
Group Lockout Boxes
Group Lockout Boxes

Group lockout centre with 18 hooks and 2 slots, Red, 406×64×195 mm, Acrylic

Price
235,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

For group lockout situations involving a large number of workers and equipment. Can also be used as a lock storage box.

  • Transparent front cover allows for easy viewing of contents
  • Hooks for keys and slot for locks
  • Components not included
Specifications
  • Colour:
    Red
  • Description:
    Group lockout centre with 18 hooks and 2 slots
  • Material:
    Acrylic
  • W×D×H:
    406×64×195 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.