Orders LinkContactUsLinkComponent
Mechanical Interval Timers
Mechanical Interval Timers
Interval timer, 60 min, with alarm, for lying, standing or hanging, Blue
Catalog # HECH41974024
Mechanical Interval Timers
Catalog # HECH41974024
Mechanical Interval Timers
Mechanical Interval Timers
Interval timer, 60 min, with alarm, for lying, standing or hanging, Blue
Price
11,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Interval timers with 60 or 120 minutes timing capacity.
  • Alarm (3 to 8 seconds)
  • Easy operation
  • For hanging, standing or lying
Specifications
  • Colour:
    Blue
  • Description:
    Interval timer, 60 min, with alarm, for lying, standing or hanging
  • Seq:
    01