Order Entry
Belgium
Orders LinkContactUsLinkComponent
Humidity standards

Humidity standards

Certified humidity standards, ±0,3% RH, 20%, 5 Ampoul
Catalog # 620-2758
Humidity standards
Catalog # 620-2758
CAS Number:  
Humidity standards
Humidity standards

Certified humidity standards, ±0,3% RH, 20%, 5 Ampoul

CAS Number:  
Price
88,00 €
Log in for availability
1 * 5 Ampul
Product Details & Documents

Although most relative humidity (RH) probes have excellent long-term stability, it is recommended that their calibration be checked regularly.

  • Traceable to national standard – SCS (Swiss Calibration Service) certificate included
  • Ampoules contain unsaturated salt solutions (~0,8 ml)
  • Inexpensive on-site calibration
  • Simple and safe to use
  • 10 year shelf life
  • Practical packs of five ampoules of the same humidity value

Delivery information: Each ampoule is labelled with the humidity value and serial number.

Caution: Values and accuracy (measurement uncertainty values) below are measured at 23 °C.

Specifications

Specifications

  • Pk:
    5 Ampoul
  • Description:
    Certified humidity standards
  • Relative humidity (RH%):
    20%
  • Accuracy:
    ±0,3% RH

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.