Orders LinkContactUsLinkComponent
Safety spectacles, 511
Safety spectacles, 511
Anti-UV, anti-scratch, anti-fog, Black, Clear PC
Catalog # UNVT511.00.03.01
Supplier:   UNIVET
Safety spectacles, 511
Catalog # UNVT511.00.03.01
Supplier:  UNIVET
Safety spectacles, 511
Safety spectacles, 511
Anti-UV, anti-scratch, anti-fog, Black, Clear PC
Price
72,20 €
Log in for availability
1 * 10 ST
Product Details & Documents
Safety spectacles, 511 has an essential design which offers good performance and the possibility of many different lenses, to meet different needs.
  • Adjustment of temples in length and inclination
  • Built-in side shields
  • Frame made of an allergic material
  • Mirrored version
These spectacles are used for mechanical works in good visibility conditions.
Wearing these spectacles for prolonged period of time will not create distortion or fatigue.
Weight: 34 g
Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    Black
  • Seq:
    02
  • Lenses:
    Clear PC