Order Entry
Belgium
Orders LinkContactUsLinkComponent
Precision butyrometer, without stopper

Precision butyrometer, without stopper

Butyrometer, precision, 0 - 4 % : 0,05, without stopper
Catalog # GERB3150-O
Supplier:   GERBER FUNKE
Precision butyrometer, without stopper
Catalog # GERB3150-O
Supplier:  GERBER FUNKE
Precision butyrometer, without stopper

Butyrometer, precision, 0 - 4 % : 0,05, without stopper

Price
126,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.

Specifications
  • Description:
    Butyrometer, precision, 0 - 4 % : 0,05, without stopper
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log in for availability
    1 * 100 ST - 48,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.