5 COMPOSES DANS HNO3 2% Na 30mg/l K 60mg/l Zn 30mg/l Fe 200mg/l Pb 200mg/l 1 * 1 L
Catalog # CMPL8B87.K2.2N.1L
Supplier: CUSTOM MADE CHEMICALS LAB
5 COMPOSES DANS HNO3 2% Na 30mg/l K 60mg/l Zn 30mg/l Fe 200mg/l Pb 200mg/l 1 * 1 L
Catalog # CMPL8B87.K2.2N.1L
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection