Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth

VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth

Grey, 1200×5400 mm, 15 mm
Catalog # 115-1199
Supplier:   VWR Collection
VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth
Catalog # 115-1199
Supplier:  VWR Collection
VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth
VWR®, Ergonomic Floor Mats, Basic Bubble and Basic Smooth

Grey, 1200×5400 mm, 15 mm

Price
845,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These mats are made of low density polyurethane providing an ergonomic, anti-fatigue profile. Bubble mats are designed for dry, non critical environments, reducing trip hazards, body aches, quality issues and sick leave. The smooth, flat mats are ideal for dry, non critical environments where sitting and standing work areas are combined, or where carts or trolleys must be used.

  • Moulded bevelled edges will not curl up, thus eliminating trip hazards
  • Any size or any shape - will always be one piece: No overlapping or worn out connecting mats
  • Ergonomic design stimulates the body to increase blood circulation and alertness
  • Silicone- and latex-free
  • Warranty up to 3 years

DIN 53505, EN ISO 527-1, ASTM D 624

Certifications: DIN 53505, EN ISO 527-1, ASTM D 624

Specifications
  • Colour:
    Grey
  • L×W:
    1200×5400 mm
  • Thickness:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.