Inlet liners
Liner, ultra inert, split, straight, glass wool TouchLess package with non-stick O-ring, 4 mm
Catalog # HEWL5190-2294
Supplier: Agilent
Jump to:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifications
- Ø int.:4 mm
- Description:Liner, ultra inert, split, straight, glass wool TouchLess package with non-stick O-ring
- Seq:19
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.