Hydrochloric acid 35%, TECHNICAL
Plastic container, 10 L
Hydrochloric acid 35%, TECHNICAL
Catalog # PRLS20246.410
CAS Number: 7647-01-0
Jump to:
Discontinued
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Boiling Pt:110 °C (1013 hPa)
- Melting Pt:–30 °C
- Density:1.18 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
- REACH:01-2119484862-27
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952503) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Specifications
Specification Test Results
- Assay:Min. 35 %
Specifications
- Pack type:Plastic container
- Pk:10 L
- Cat. No.:PRLS20246.410
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



