Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen monitor, dor detecting trace oxygen levels using an electrochemical sensor; digital readout in three measurement ranges of 0-100 ppm, 100-1000 ppm and 1000-10000 ppm, for 230 V units
Catalog # LBCP5244501
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5244501
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
Oxygen monitor, dor detecting trace oxygen levels using an electrochemical sensor; digital readout in three measurement ranges of 0-100 ppm, 100-1000 ppm and 1000-10000 ppm, for 230 V units
Price
10 800,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Oxygen monitor, dor detecting trace oxygen levels using an electrochemical sensor; digital readout in three measurement ranges of 0-100 ppm, 100-1000 ppm and 1000-10000 ppm, for 230 V units
  • Seq:
    16