Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Liner, ultra inert deactivation, splitless, single taper, glass wool, 4 mm
Catalog # HEWL5190-3167
Supplier:   Agilent
Inlet liners
Catalog # HEWL5190-3167
Supplier:  Agilent
Inlet liners
Inlet liners
Liner, ultra inert deactivation, splitless, single taper, glass wool, 4 mm
Price
971,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specifications
  • Ø int.:
    4 mm
  • Description:
    Liner, ultra inert deactivation, splitless, single taper, glass wool
  • Seq:
    07
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.