ContactUsLinkComponent
15 COMPOSES DANS ACETONITRILE ( ampoules de 1 ml ) Naphthalene [CAS:91-20-3] 500mg/l Acenaphthene [CAS:83-32-9] 1000mg/l Fluorene [CAS:86-73-7] 100mg/l Phenanthrene [CAS:85-01-8] 40mg/l Anthracene [CAS:120-12-7] 20mg/l Fluoranthene [CAS:206 1 * 5 Ampoul
Catalog # CMPL06F8.20.N.1
15 COMPOSES DANS ACETONITRILE ( ampoules de 1 ml ) Naphthalene [CAS:91-20-3] 500mg/l Acenaphthene [CAS:83-32-9] 1000mg/l Fluorene [CAS:86-73-7] 100mg/l Phenanthrene [CAS:85-01-8] 40mg/l Anthracene [CAS:120-12-7] 20mg/l Fluoranthene [CAS:206 1 * 5 Ampoul
Catalog # CMPL06F8.20.N.1
Frequently Bought Together