Order Entry
Belgium
Orders LinkContactUsLinkComponent
Syringes for CTC analytics systems

Syringes for CTC analytics systems

Removable needle, 23, 2 µl, 50 mm, 0,63 mm, Conical, 2BR-C/T-5/0.63C
  SGEA000790
Syringes for CTC analytics systems
  SGEA000790
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Removable needle, 23, 2 µl, 50 mm, 0,63 mm, Conical, 2BR-C/T-5/0.63C

 
153,00 €
   
1 * 1 ST
 
  • Ext.Ø:
    0,63 mm
  • Gauge:
    23
  • Type:
    2BR-C/T-5/0.63C
  • Type of tip:
    Conical
  • Volume:
    2 µl
  • Description:
    Removable needle
  • Length:
    50 mm
  • Seq:
    17
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.