Order Entry
Belgium
Orders LinkContactUsLinkComponent
Container, rectangular, with handle

Container, rectangular, with handle

6 l, 198×290×169 mm, -
Catalog # 216-1607
Supplier:   HK PLASTICS
Container, rectangular, with handle
Catalog # 216-1607
Supplier:  HK PLASTICS
Container, rectangular, with handle
Container, rectangular, with handle

6 l, 198×290×169 mm, -

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

PP/PE.

  • Increase storage capacity, reduce the transport volume and the required shelf space
  • All materials are food grade approved and coloured master batch, cadmium free
  • Designed to be nestable and interlocking

Robust rectangular containers, which are easy to open and (re)close. Strong, ergonomic handle grips making them easy to transport.

Produced under BRC/IoP regulation, according to HACCP standards.

Specifications
  • Type:
    -
  • Capacity:
    6 l
  • W×D×H:
    198×290×169 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.