Orders LinkContactUsLinkComponent
Separating funnel, globe shaped
Separating funnel, globe shaped
250 ml, 29/32 NS
Catalog # KAVA632426351250
Supplier:   Kavalier
Separating funnel, globe shaped
Catalog # KAVA632426351250
Supplier:  Kavalier
Separating funnel, globe shaped
Separating funnel, globe shaped
250 ml, 29/32 NS
Price
314,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for separation of particular phases of the mixture. With standard ground joint stopper.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml