Test kits, digital titration method
Mercuric nitrate* 2,256 N, 13 mL
Catalog # HACH92101
Supplier: Hach
Aller à :
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Attention: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spécifications
Spécifications
- Pk:13 mL
- Description:Mercuric nitrate* 2,256 N



