Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with Quickfit® ground joint

Flasks, Erlenmeyer, with Quickfit® ground joint

1000 ml, 220 mm, 130 mm, 29/32 NS
Catalog # BIBBFE1L/4
Supplier:   SciLabware
Flasks, Erlenmeyer, with Quickfit® ground joint
Catalog # BIBBFE1L/4
Supplier:  SciLabware
Flasks, Erlenmeyer, with Quickfit® ground joint
Flasks, Erlenmeyer, with Quickfit® ground joint

1000 ml, 220 mm, 130 mm, 29/32 NS

Price
286,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

PYREX® borosilicate glass, clear. With matt white labelling area.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

ISO 4797

Certifications: ISO 4797

Specifications
  • Ext.Ø:
    130 mm
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Height:
    220 mm
  • Seq:
    24
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.