Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fractionating columns, plain, Quickfit®

Fractionating columns, plain, Quickfit®

500 mm, 615 mm, 24/29 NS, 24/29 NS
Catalog # BIBBFC7/43
Supplier:   SciLabware
Fractionating columns, plain, Quickfit®
Catalog # BIBBFC7/43
Supplier:  SciLabware
Fractionating columns, plain, Quickfit®
Fractionating columns, plain, Quickfit®

500 mm, 615 mm, 24/29 NS, 24/29 NS

Price
55,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

PYREX®, borosilicate glass

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Ground glass joints for dependable performance. Designed for use with various types of column packing e.g. Raschig rings or Fenske helices.

Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Usable length:
    500 mm
  • Seq:
    02
  • Total length:
    615 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.