Order Entry
Belgium
Orders LinkContactUsLinkComponent
PIG® Essentials Universal Socks

PIG® Essentials Universal Socks

PIG® essentials universal socks, 94,6 L/pack, Grey, 80×1220 mm
Catalog # NEWPMS430
Supplier:   NEW PIG
PIG® Essentials Universal Socks
Catalog # NEWPMS430
Supplier:  NEW PIG
PIG® Essentials Universal Socks
PIG® Essentials Universal Socks

PIG® essentials universal socks, 94,6 L/pack, Grey, 80×1220 mm

Price
109,00 €
Log in for availability
1 * 30 ST
Product Details & Documents

Polypropylene-filled socks and booms to absorb oils, coolants, solvents and water.

  • 100% polypropylene-filled socks and booms absorb oils, coolants, solvents and water
  • Ideal for surrounding spilled fluids or wrapping around leaky machines
  • Easy and convenient to put into place; contain spills to keep fluids from spreading into larger areas and becoming slippery hazards
  • Flexible to conform around machine bases and offer a snug fit with no seepage
  • Grey colour signifies Universal sorbency for easy visual identification
  • Contains 100% post-industrial recycled poly blend filler

The product consists of a polypropylene absorbent. Contained within a casing made of: Polypropylene (CAS 9003-07-0). Also containing impregnated: Surfactant. Pigment. May contain: Mixture Adhesive; Polypropylene film; Polyester yarn.

Specifications
  • Colour:
    Grey
  • Description:
    PIG® essentials universal socks
  • Seq:
    01
  • Absorbency:
    94,6 L/pack
  • Ø×L:
    80×1220 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.