Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Manual cell counters

Manual cell counter, 10
  MOIN1495/2MW
 :   MOINEAU
Manual cell counters
  MOIN1495/2MW
 :  MOINEAU
Manual cell counters
Manual cell counters

Manual cell counter, 10

 
793,00 €
   
1 * 1 ST
 

This manual cell counter can be used in combination of up to five counters.

  • Four digit display (4,3×3,5 mm) with reset knob
  • Use finger pressure
  • Can be fixed in position

W×D×H: 37,6×0,4×38,1 mm

 
  • Description:
    Manual cell counter
  • Seq:
    005
  • Array:
    10