PAPER FOR PRINTER EPSON-TM-U220 1 * 5 Roll
Catalog # MBVA04.4302.01
Supplier: MBV
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Laboratory Bottles, Wide Neck
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection