Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
25×60 mm, 1,0 mm
Catalog # WHAT10350416
Extraction thimbles
Catalog # WHAT10350416
Supplier:  
Extraction thimbles
Extraction thimbles
25×60 mm, 1,0 mm
Discontinued
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Specifications
  • Int.Ø×H:
    25×60 mm
  • Seq:
    01
  • Thickness:
    1,0 mm