Filter vials, SINGLE StEP™
PES, 0,2 µm, Grey
Catalog # 548-1405
Supplier: THOMSON INSTRUMENT
Jump to:
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
- Compatible with both UHPLC and standard HPLC autosamplers
- Colour coded pre-slit caps for easy identification of membrane and porosity
- Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Specifications
- Membrane:PES
- Pore size:0,2 µm
- Colour:Grey
- Seq:03