Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pyridine, extra pure

Pyridine, extra pure

Glass bottle, 500 mL
Catalog # KRAF07051.2700
Flammable
Irritant
Supplier:   Bernd Kraft
Pyridine, extra pure
Catalog # KRAF07051.2700
Supplier:  Bernd Kraft
CAS Number:  110-86-1
Pyridine, extra pure
Pyridine, extra pure

Glass bottle, 500 mL

CAS Number:  110-86-1
Price
75,20 €
Log in for availability
1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.