Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solution pH 10

Buffer solution pH 10

Buffer solution, 10, Ammonia, 10 L
Catalog # KRAF03596.6000
Supplier:   Bernd Kraft
Buffer solution pH 10
Catalog # KRAF03596.6000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution pH 10

Buffer solution, 10, Ammonia, 10 L

CAS Number:  
Price
310,00 €
Log in for availability
1 * 10 L
Product Details & Documents

This solution is a combination of 53,5 g NH₄Cl, 350 ml NH₃ solution 25% and Cu-EDTA/l used for complexometric determination of zinc.

Specifications

Specifications

  • pH buffer type:
    Ammonia
  • Pk:
    10 L
  • Description:
    Buffer solution
  • pH value:
    10

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.