Water
Plastic bottle, 2.500 mL
Catalog # KRAF10791.4000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:2.500 mL



