Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

Plastic bottle, 0,5 L
Catalog # KRAF10791.2000
Supplier:   Bernd Kraft
Water
Catalog # KRAF10791.2000
Supplier:  Bernd Kraft
CAS Number:  7732-18-5
Water

Plastic bottle, 0,5 L

CAS Number:  7732-18-5
Price
Inquire For Price
Log in for availability
1 * 0,5 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    0,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.