Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

4,66, Acetate, Colourless, 20 °C, Plastic bottle, 10 L
  KRAF03397.6400
Buffer solutions, ready to use
  KRAF03397.6400
 :  
Buffer solutions, ready to use
Buffer solutions, ready to use

4,66, Acetate, Colourless, 20 °C, Plastic bottle, 10 L

 :  
 
196,00 €
   
1 * 10 L
 
  • NIST traceable

 

 

 

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Acetate
  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    4,66