Buffer solution pH 1,09
Buffer solution, 1,09 (25 °C), 10 L
Catalog # KRAF03853.6400
Supplier: Bernd Kraft
Aller à :
Product Details & Documents
- Stabilized
: According to DIN 19267.
Spécifications
Spécifications
- Pk:10 L
- Description:Buffer solution
- pH value:1,09 (25 °C)



