Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solution pH 1,09

Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 5 L
Catalog # KRAF03853.5400
Supplier:   Bernd Kraft
Buffer solution pH 1,09
Catalog # KRAF03853.5400
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution pH 1,09
Buffer solution pH 1,09

Buffer solution, 1,09 (25 °C), 5 L

CAS Number:  
Price
221,00 €
Connexion Pour la disponibilité
1 * 5 L
Product Details & Documents
  • Stabilized

Certifications: According to DIN 19267.

Spécifications

Spécifications

  • Pk:
    5 L
  • Description:
    Buffer solution
  • pH value:
    1,09 (25 °C)

Spécifications