SnCl * 2 HO in verdunnter Salzsaure zur Quecksilber-Bestimmung gemaß EN 1483:2007. DEV E12 1 * 1 L
Catalog # KRAF16858.3100
Supplier: Bernd Kraft
SnCl * 2 HO in verdunnter Salzsaure zur Quecksilber-Bestimmung gemaß EN 1483:2007. DEV E12 1 * 1 L
Catalog # KRAF16858.3100
Supplier: Bernd Kraft
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
pH buffers, ready to use
Supplier: WTW
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals