Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium iodide 30% in aqueous solution analytical reagent

Potassium iodide 30% in aqueous solution analytical reagent

Glass bottle, 500 mL
Catalog # KRAF03327.2700
Supplier:   Bernd Kraft
Potassium iodide 30% in aqueous solution analytical reagent
Catalog # KRAF03327.2700
Supplier:  Bernd Kraft
CAS Number:  7681-11-0
Potassium iodide 30% in aqueous solution analytical reagent

Glass bottle, 500 mL

CAS Number:  7681-11-0
Price
110,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-659-4

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications